2-(Methylthio)naphtho[1,2-d]thiazole ≥98.0% (by GC)
Catalog # TCIAM1827-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Purity (GC):min. 98.0 %
- Melting point:62.0 to 66.0 deg-C
Specification Test Results
About this item
:
:231.33 g/mol
Boiling Pt:248°C
CAS Number:51769-43-8
Formula:C₁₂H₉NS₂