Order Entry
Sweden
ContactUsLinkComponent
4-Methoxyphenyl-2-azido-4,6-O-benzylidene-2-deoxy-β-D-glucopyranoside ≥98.0% (by HPLC)
4-Methoxyphenyl-2-azido-4,6-O-benzylidene-2-deoxy-β-D-glucopyranoside ≥98.0% (by HPLC)
Supplier:  TCI
Certificates
4-Methoxyphenyl-2-azido-4,6-O-benzylidene-2-deoxy-β-D-glucopyranoside ≥98.0% (by HPLC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Melting point:
    160.0 to 165.0 deg-C
  • Specific rotation [a]20/D:
    -3.0 to -6.0 deg (C=1, CHCl3)
  • Solubility in Chloroform:
    very faint turbidity

Specification Test Results

Frequently Bought Together