4-Methoxyphenyl-3-O-allyl-4,6-O-benzylidene-2-deoxy-2-phthalimido-β-D-glucopyranoside ≥97.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:889453-84-3
:
Formula:C₃₁H₂₉NO₈
:543.57 g/mol
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 97.0 area%
- Melting point:128.0 to 132.0 deg-C
- Specific rotation [a]20/D:+30.0 to +36.0 deg (C=1,CHCl3)
Specification Test Results
Frequently Bought Together