Methyl-2,3,4,6-tetra-O-acetyl-1-thio-ɑ-D-mannopyranoside ≥95.0% (by HPLC) (contains ca. 5% beta-isomer)
Catalog # TCIAM1501-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 95.0 area%
- Melting point:123.0 to 127.0 deg-C
- Specific rotation [a]20/D:+90.0 to +98.0 deg (C=1,chloroform)
Specification Test Results
About this item
:
:378.39 g/mol
Storage Temperature:Freezer
CAS Number:64550-71-6
Formula:C₁₅H₂₂O₉S
MDL Number:MFCD00080808