4-Methoxyphenyl-3,4,6-tri-O-acetyl-2-deoxy-2-phthalimido-β-D-glucopyranoside ≥98.0% (by HPLC)
TCIAM1480-5G
: TCI
:
- Pk:5 g
- Seq:3173
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (HPLC):min. 98.0 area%
- Melting point:146.0 to 150.0 deg-C
- Specific rotation [a]20/D:+52.0 to +56.0 deg (C=1, CHCl3)
- Solubility in Chloroform:almost transparency
:
:541.51 g/mol
:Freezer
:138906-41-9
:C₂₇H₂₇NO₁₁
:MFCD06797125