4-Methoxyphenyl-4,6-O-benzylidene-2-deoxy-2-phthalimido-β-D-glucopyranoside ≥98.0% (by HPLC)
Catalog # TCIAM1479-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Melting point:136.0 to 142.0 deg-C
- Specific rotation [a]20/D:+10.0 to +13.0 deg (C=1, CHCl3)
- Solubility in Chloroform:very faint turbidity
Specification Test Results
About this item
:
:503.51 g/mol
Storage Temperature:Freezer
CAS Number:138906-43-1
Formula:C₂₈H₂₅NO₈
MDL Number:MFCD06797124