Order Entry
Sweden
ContactUsLinkComponent
N-Methyl-DL-alanine ≥98.0% (by titrimetric analysis)
N-Methyl-DL-alanine ≥98.0% (by titrimetric analysis)
Catalog # TCIAM0777-100MG
Supplier:  TCI
CAS Number:  
N-Methyl-DL-alanine ≥98.0% (by titrimetric analysis)
Catalog # TCIAM0777-100MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (Neutralization titration):
    min. 98.0 %

Specification Test Results

About this item

 :
 :103.12 g/mol
Melting Pt:317°C
CAS Number:600-21-5
Formula:C₄H₉NO₂
MDL Number:MFCD00063136