Order Entry
Sweden
ContactUsLinkComponent
 
 
 
 
 
3-Methyl-2-butenal ≥97.0% (by GC)
 :  TCI
 
undefined
 
 
 
 
3-Methyl-2-butenal ≥97.0% (by GC)
 :  TCI

 

  • Appearance:
    Colorless to Light orange to Yellow clear liquid
  • Purity (GC):
    min. 97.0 %