L-Methionylglycine
Catalog # TCIAM0250-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- CAS No.:14486-03-4
- Form:White to almost white powder to crystal
- MDL No.:MFCD00021730
- Protein synonyms:H-Met-Gly-OH
- Protein/peptide name:L-Methionylglycine
- Purity:Min. 98,0% (HPLC)
- Molecular weight:206,26 g/mol
- Molecular formula:C₇H₁₄N₂O₃S
Specifications
About this item
- Melting point: 212 °C
- Purity (Nonaqueous titration): Min. 95.0%
- Specific rotation [a]20/D: 85.0 to 90.0° (C=2, Water)
- Solubility in water: almost transparency
- EINECS: 238-487-9
Other Product Names / Synonyms: H-Met-Gly-OH