(3-Indoxyl)-β-D-glucopyranoside ≥98.0% (by HPLC)
Catalog # TCIAI1012-200MG
Supplier: TCI
CAS Number:
Specifications
- Pk:200 mg
- Seq:2530
Specifications
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 97.0 %
- Specific rotation [a]20/D:-62.0 to -67.0 deg (C=2, H2O)
Specification Test Results
About this item
:
Merck Index:14,04942
Storage Temperature:Freezer
CAS Number:487-60-5
Formula:C₁₄H₁₇NO₆
MDL Number:MFCD00047169