Order Entry
Sweden
ContactUsLinkComponent
N2-Isobutyryl-5'-O-(4,4'-dimethoxytrityl)-2'-deoxyguanosine ≥99.0% (by HPLC)
N2-Isobutyryl-5'-O-(4,4'-dimethoxytrityl)-2'-deoxyguanosine ≥99.0% (by HPLC)
Supplier:  TCI
Certificates
N2-Isobutyryl-5'-O-(4,4'-dimethoxytrityl)-2'-deoxyguanosine ≥99.0% (by HPLC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White powder to crystal
  • Purity (HPLC):
    min. 99.0 area%
  • Specific rotation [a]20/D:
    +8.0 to +9.5deg (C=1, methanol)
  • Water:
    max. 3.0 %

Specification Test Results

Related Information