
4-Hydroxy-7-methoxycoumarin ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:17575-15-4
:
Formula:C₁₀H₈O₄
MDL Number:MFCD00673700
:192.17 g/mol
:GHS07
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %