Order Entry
Sweden
ContactUsLinkComponent
bisBenzimide H 33258 trihydrochloride hydrate (Hoechst 33258) ≥98.0% (by HPLC) fluorescent dye, for biochemical research
Irritant
bisBenzimide H 33258 trihydrochloride hydrate (Hoechst 33258) ≥98.0% (by HPLC) fluorescent dye, for biochemical research
Catalog # TCIAH1343-25MG
Supplier:  TCI
CAS Number:  
undefined
Irritant
bisBenzimide H 33258 trihydrochloride hydrate (Hoechst 33258) ≥98.0% (by HPLC) fluorescent dye, for biochemical research
Catalog # TCIAH1343-25MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    25 mg
  • Seq:
    2315

Specifications

Specification Test Results

  • Appearance:
    Light yellow to Brown to Dark green powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Water (Vaporization methoed):
    max. 20.0 %
  • DNA staining test:
    to pass test

Specification Test Results

About this item

 :
 :533.88 g/mol (anhy)
Melting Pt:280°C
EINECS:245-690-6
 :GHS07
Storage Temperature:Freezer
CAS Number:23491-45-4
Formula:C₂₅H₂₄N₆O·3HCl·xH₂O
MDL Number:MFCD00012679