Order Entry
Sweden
ContactUsLinkComponent
 
 
6-Hydroxy-1-indanone ≥98.0% (by GC)
 :  TCI
 
undefined
 
6-Hydroxy-1-indanone ≥98.0% (by GC)
 :  TCI

 

  • Appearance:
    Light yellow to Amber to Dark green powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    155.0 to 159.0 deg-C

 

Frequently Bought Together