Order Entry
Sweden
ContactUsLinkComponent
 
Potassium-1,1,2,2,3,3-hexafluoropropane-1,3-disulphonimide ≥98.0% (by titrimetric analysis)
 :  TCI
 
Potassium-1,1,2,2,3,3-hexafluoropropane-1,3-disulphonimide ≥98.0% (by titrimetric analysis)
 :  TCI

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (Ion exchange titration):
    min. 98.0 %
  • Solubility in Water:
    almost transparency