(Hexamethylbenzene)ruthenium(II) dichloride ≥95.0% (by titrimetric analysis)
Catalog # TCIAH1010-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:Orange to Brown powder to crystal
- Purity (Argentmetric Titration):min. 95.0 %
Specification Test Results
About this item
:
:668.49 g/mol
CAS Number:67421-02-7
Formula:C₂₄H₃₆Cl₄Ru₂
MDL Number:MFCD03701102