Order Entry
Sweden
ContactUsLinkComponent
Dichlorohexylmethylsilane ≥95.0% (by GC)
Corrosive
Dichlorohexylmethylsilane ≥95.0% (by GC)
Catalog # TCIAH0855-25ML
Supplier:  TCI
CAS Number:  
Corrosive
Dichlorohexylmethylsilane ≥95.0% (by GC)
Catalog # TCIAH0855-25ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    25 mL
  • Seq:
    2315

Specifications

Specification Test Results

  • Appearance:
    Colorless to Light yellow to Light orange clear liquid
  • Purity (GC):
    min. 95.0 %

Specification Test Results

About this item

 :
 :199.19 g/mol
Boiling Pt:103°C
Flash Pt:85°C
EINECS:238-864-8
UN:2987
ADR:8,II
 :GHS05
CAS Number:14799-94-1
Formula:C₇H₁₆Cl₂Si
MDL Number:MFCD00039295