ɑ-Hydroxy-γ-butyrolactone ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:19444-84-9
:
Formula:C₄H₆O₃
MDL Number:MFCD00134268
:102.09 g/mol
Boiling Pt:133°C
Specification Test Results
- Appearance:Colorless to Red to Green clear liquid
- Purity (GC):min. 98.0 %