Tris(2,4-dimethoxyphenyl)methanol
Catalog # TCIAH0742-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Melting point:150.0 to 154.0 deg-C
- Absorbance (E1%1cm):min. 700 (20mg/L, 1mol/L HCl, 550.0 to 553.0 nm)
- Transition interval (pH):(red)2.6 ~ 4.6 (slightly pale red)
Specification Test Results
About this item
:
:440.49 g/mol
CAS Number:76832-37-6
Formula:C₂₅H₂₈O₇
MDL Number:MFCD00145468