Order Entry
Sweden
ContactUsLinkComponent
DL-4-Hydroxymandelic acid monohydrate ≥98.0% (by HPLC, titration analysis)
Irritant
DL-4-Hydroxymandelic acid monohydrate ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAH0660-25G
Supplier:  TCI
CAS Number:  
Irritant
DL-4-Hydroxymandelic acid monohydrate ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAH0660-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Neutralization titration):
    min. 98.0 % (calcd.on anh.substance)
  • Water:
    9.2 to 10.2 %

Specification Test Results

About this item

 :
 :186.16 g/mol
EINECS:230-569-2
 :GHS07
CAS Number:184901-84-6
Formula:C₈H₁₀O₅
MDL Number:MFCD00149283