Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Proteins and Peptides
Glycyl-L-Aspartic Acid Monohydrate
Glycyl-L-Aspartic Acid Monohydrate
Glycyl-L-Aspartic Acid Monohydrate
TCIAG0121-1G
:
TCI
:
Glycyl-L-Aspartic Acid Monohydrate
TCIAG0121-1G
:
TCI
:
Pk:
1 g
CAS No.:
4685-12-5
Protein synonyms:
Glycyl-L-aspartic acid monohydrate
Protein/peptide name:
H-Gly-Asp-OH
Purity:
≥98.0% (by titrimetric analysis)