Nalpha-Glycyl-L-asparagine
Catalog # TCIAG0118-1G
Supplier: TCI
CAS Number:
Specifications
- Förp.:1 g
- CAS-nr:1999-33-3
- Form:White to almost white powder to crystal
- MDL No.:MFCD00066024
- Protein synonyms:H-Gly-Asn-OH
- Protein/peptide name:Nɑ-Glycyl-L-asparagine
- Renhet:Min. 98,0% (nonaqueous titration)
- Molekylvikt:189,17 g/mol
- Molekylformel:C₆H₁₁N₃O₄
Specifications
About this item
- Specific rotation [a]20/D: –8° (C=1, H₂O)
- Solubility in water: almost transparency
- EINECS: 217-881-4
Product Categories: Bioscience, Oligopeptides
Other Product Names / Synonyms: H-Gly-Asn-OH