6-FAM-PEG3-Azide ≥95.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:412319-45-0
:
Formula:C₂₉H₂₈N₄O₉
:576.56 g/mol
Storage Temperature:Freezer
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 95.0 area%