
N-(4-Fluorophenyl)glycine ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:351-95-1
:
Formula:C₈H₈FNO₂
MDL Number:MFCD02255629
:169.16 g/mol
Melting Pt:138°C
:GHS07
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %