
2-Fluoro-4-pyridineboronic acid (contains varying amounts of Anhydride)
Supplier: TCI
Certificates
About this item
CAS Number:401815-98-3
:
Formula:C₅H₅BFNO₂
MDL Number:MFCD04112345
:140.91 g/mol
Melting Pt:200°C
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Neutralization titration):98.0 to 115.0 %