Order Entry
Sweden
ContactUsLinkComponent
4-Ethynyl-1,2-difluorobenzene ≥98.0% (by GC)
Flammable
Irritant
4-Ethynyl-1,2-difluorobenzene ≥98.0% (by GC)
Catalog # TCIAE1349-1G
Supplier:  TCI
CAS Number:  
Flammable
Irritant
4-Ethynyl-1,2-difluorobenzene ≥98.0% (by GC)
Catalog # TCIAE1349-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 98.0 %

Specification Test Results

About this item

 :
 :138.12 g/mol
Flash Pt:7°C
UN:1993
ADR:3,II
 :GHS02
 :GHS07
CAS Number:143874-13-9
Formula:C₈H₄F₂
MDL Number:MFCD08059511