0,00 kr
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Sweden
ContactUsLinkComponent
4-Ethynyl-4'-propylbiphenyl
4-Ethynyl-4'-propylbiphenyl ≥98.0% (by GC)
4-Ethynyl-4'-propylbiphenyl ≥98.0% (by GC)
:
TCI
4-Ethynyl-4'-propylbiphenyl ≥98.0% (by GC)
:
TCI
:
360768-57-6
:
:
C₁₇H₁₆
:
MFCD18072535
:
220.32 g/mol
Appearance:
White to Orange to Green powder to crystal
Purity (GC):
min. 98.0 %