1-Ethynyl-4-(trifluoromethoxy)benzene ≥97.0% (by GC)
Leverantör: TCI
Certifikat
Om denna produkt
CAS-nummer:160542-02-9
:
Formel:C₉H₅F₃O
MDL-nummer:MFCD03094333
:186.13 g/mol
Flash Pt:40°C
FN:1993
ADR:3,III
:GHS02
:GHS07
Specifikationstestresultat
- Appearance:White to Yellow to Green clear liquid
- Purity (GC):min. 97.0 %