2-Ethyl-5-nitroaniline ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:20191-74-6
:
Formula:C₈H₁₀N₂O₂
MDL Number:MFCD00034056
:166.18 g/mol
:GHS07
Specification Test Results
- Appearance:Light yellow to Brown powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 96.0 %
- Melting point:58.0 to 62.0 deg-C