
4-Ethynylpyridine hydrochloride ≥95.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:352530-29-1
:
Formula:C₇H₆ClN
MDL Number:MFCD03093912
:139.58 g/mol
:GHS07
Specification Test Results
- Appearance:Yellow to Brown to Dark green powder to crystal
- Purity (HPLC):min. 95.0 area%
- Purity (Nonaqueous Titration):min. 95.0 %