Ethyl-2-oxocyclopentaneacetate ≥95.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:20826-94-2
:
Formula:C₉H₁₄O₃
MDL Number:MFCD00044715
:170.21 g/mol
Boiling Pt:106°C
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 95.0 %