Dimethyl Icosanedioate ≥95.0% (by GC)
Leverantör: TCI
Certifikat
Om denna produkt
CAS-nummer:42235-38-1
:
Formel:C₂₂H₄₂O₄
MDL-nummer:MFCD00059302
:370.57 g/mol
Kokande Pt:223°C
Specifikationstestresultat
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 95.0 %
- Melting point:65.0 to 67.0 deg-C