



3-Ethyl-2-methylpentane ≥99.0% (by GC)
Catalog # TCIAE0259-5ML
Supplier: TCI
CAS Number:
Specifications
- Pk:5 mL
- Seq:2210
Specifications
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 99.0 %
Specification Test Results
About this item
:
:114.23 g/mol
Boiling Pt:116°C
Flash Pt:3°C
EINECS:210-187-2
UN:1262
ADR:3,II
:GHS02
:GHS07
:GHS08
:GHS09
CAS Number:609-26-7
Formula:C₈H₁₈
MDL Number:MFCD00048665