
1,4-Diaminobutane dihydroiodide ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:916849-52-0
:
Formula:C₄H₁₂N₂·₂HI
:343.98 g/mol
:GHS07
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (Nonaqueous Titration):min. 98.0 %