trans,trans-3,4-Difluoro-4'-(4'-propylbicyclohexyl-4-yl)biphenyl ≥98.0% (by GC)
Leverantör: TCI
Certifikat
Om denna produkt
CAS-nummer:119990-81-7
:
Formel:C₂₇H₃₄F₂
MDL-nummer:MFCD11053440
:396.57 g/mol
Specifikationstestresultat
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %