Order Entry
Sweden
ContactUsLinkComponent
4-(N,N-Diphenylamino)phenylboronic acid pinacol ester ≥98.0% (by HPLC, titration analysis)
Irritant
4-(N,N-Diphenylamino)phenylboronic acid pinacol ester ≥98.0% (by HPLC, titration analysis)
Supplier:  TCI
Certificates
Irritant
4-(N,N-Diphenylamino)phenylboronic acid pinacol ester ≥98.0% (by HPLC, titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Neutralization titration):
    min. 98.0 %
  • Melting point:
    93.0 to 97.0 deg-C

Specification Test Results