Order Entry
Sweden
ContactUsLinkComponent
Dansyl amide ≥98.0% (by titrimetric analysis) for albumin binding assay
Dansyl amide ≥98.0% (by titrimetric analysis) for albumin binding assay
Supplier:  TCI
Certificates
undefined
Dansyl amide ≥98.0% (by titrimetric analysis) for albumin binding assay
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Melting point:
    218.0 to 222.0 deg-C
  • Human serum albmin (HSA) binding activity:
    S/N ratio min. 4
  • Competitive avtivity in competitive drug:
    Remaining fluorescein inensity max. 70 %

Specification Test Results