Order Entry
Sweden
ContactUsLinkComponent
 
2,6-Dihydroxyanthracene ≥95.0% (by GC)
 :  TCI
 
undefined
2,6-Dihydroxyanthracene ≥95.0% (by GC)
 :  TCI

 

  • Appearance:
    Light yellow to Brown powder to crystal
  • Purity (GC):
    min. 95.0 %