
Dimethylammonium bromide ≥98.0% (by total nitrogen and titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:6912-12-5
:
Formula:C₂H₈BrN
MDL Number:MFCD00051983
:126
EINECS:230-019-1
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (with Total Nitrogen):min. 98.0 %
- Purity (Argentmetric Titration):min. 98.0 %