Order Entry
Sweden
ContactUsLinkComponent
L-Glutamic acid dibenzyl ester hydrochloride ≥98.0% (by HPLC, titration analysis)
L-Glutamic acid dibenzyl ester hydrochloride ≥98.0% (by HPLC, titration analysis)
Supplier:  TCI
Certificates
undefined
L-Glutamic acid dibenzyl ester hydrochloride ≥98.0% (by HPLC, titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Melting point:
    102.0 to 106.0 deg-C
  • Specific rotation [a]20/D:
    +8.5 to +10.0 deg (C=2, H2O)

Specification Test Results