4-(3,4-Dichlorophenyl)-1-tetralone ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:79560-19-3
:
Formula:C₁₆H₁₂Cl₂O
MDL Number:MFCD02093088
:291.18 g/mol
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:104.0 to 108.0 deg-C