
2 3-Dichloro-6-nitroquinoxaline ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:2379-60-4
:
Formula:C₈H₃Cl₂N₃O₂
MDL Number:MFCD00454905
:244.0343 g/mol
:GHS07
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:151.0 to 155.0 deg-C