Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Diethyl propylmalonate
Diethyl propylmalonate ≥98.0% (by GC)
Diethyl propylmalonate ≥98.0% (by GC)
:
TCI
Diethyl propylmalonate ≥98.0% (by GC)
:
TCI
:
2163-48-6
:
:
C₁₀H₁₈O₄
:
MFCD00009168
:
202.25 g/mol
:
222°C
:
92°C
Appearance:
Colorless to Almost colorless clear liquid
Purity (GC):
min. 98.0 %