
3',5'-Dibromo-2'-hydroxyacetophenone ≥98.0% (by GC, titration analysis)
: TCI
:22362-66-9
:
:C₈H₆Br₂O₂
:MFCD00191992
:293.95 g/mol
:GHS07
- Appearance:White to Yellow to Orange powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:108.0 to 112.0 deg-C