Doxepin hydrochloride ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:1229-29-4
:
Formula:C₁₉H₂₂ClNO
MDL Number:MFCD00079135
:315.841 g/mol
Merck Index:14,03435
UN:2811
ADR:6.1,III
:GHS06
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 % (total of isomers)
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:186.0 to 190.0 deg-C