Order Entry
Sweden
ContactUsLinkComponent
 
4-(Dicyanomethylene)-2-methyl-6-[2-(2,3,6,7-tetrahydro-1H,5H-benzo[ij]quinolizin-9-yl)vinyl]-4H-pyran ≥97.0% (by HPLC, total nitrogen)
Catalog # TCIAD4459-200MG
Supplier:  TCI
CAS Number:  
4-(Dicyanomethylene)-2-methyl-6-[2-(2,3,6,7-tetrahydro-1H,5H-benzo[ij]quinolizin-9-yl)vinyl]-4H-pyran ≥97.0% (by HPLC, total nitrogen)
Catalog # TCIAD4459-200MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    200 mg
  • Seq:
    2530

Specifications

Specification Test Results

  • Purity (HPLC):
    min. 97.0 area%
  • Purity (with Total Nitrogen):
    min. 97.0 %
  • Lambda max.:
    505.0 to 510.0 nm (CH2Cl2)
  • Molar extinction coefficient:
    min. 43000 (CH2Cl2, 505.0 to 510.0nm)

Specification Test Results

About this item

 :
 :355.44 g/mol
 :GHS07
CAS Number:51325-95-2
Formula:C₂₃H₂₁N₃O
MDL Number:MFCD02249183