
2,2-Dimethyl-4-pentenoic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:16386-93-9
:
Formula:C₇H₁₂O₂
MDL Number:MFCD00012304
:128.17 g/mol
Boiling Pt:108°C
Flash Pt:88°C
UN:3265
ADR:8,II
:GHS05
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %