

2,2-Diphenyl-1-picrylhydrazyl (free radical) ≥97.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:1898-66-4
:
Formula:C₁₈H₁₂N₅O₆
MDL Number:MFCD00007231
:394.32 g/mol
Merck Index:14,03330
EINECS:217-591-8
:GHS07
:GHS08
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Black powder to crystal
- Purity (HPLC):min. 97.0 area%