DAPT ≥97.0% (by HPLC)
Catalog # TCIAD4257-25MG
Supplier: TCI
CAS Number:
Specifications
- Pk:25 mg
- Seq:2315
Specifications
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (HPLC):min. 97.0 area%
- Elemental analysis (Nitrogen):6.20 to 6.70 %
Specification Test Results
About this item
:
:432.47 g/mol
Storage Temperature:Refrigerator
CAS Number:208255-80-5
Formula:C₂₃H₂₆F₂N₂O₄
MDL Number:MFCD04974585