4-(1,1-Dioxothiomorpholino)benzaldehyde ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:27913-96-8
:
Formula:C₁₁H₁₃NO₃S
MDL Number:MFCD08276340
:239.29 g/mol
Specification Test Results
- Appearance:White to Yellow to Green powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:183.0 to 187.0 deg-C
- Solubility in hot Acetone:almost transparency